Learn More
Biosynergy TempTrend™ II Forehead Core Temperature Indicators
Allows continuous temperature monitoring of all patients
Manufacturer:  Biosynergy 1100B
Catalog No. 22-246192
Add to cart
Description
Description
  • Disposable, liquid-crystal forehead indicators gives noninvasive alternative to electronic temperature probes
  • Adjusted to read core temperature as measured by oral thermometer
  • Postage stamp sized indicators provides specific, single-spot reading
  • Can be placed on patients before therapy and can be retained during and after therapy
  • Results in 1 to 15 seconds
Specifications
Specifications
Measures body temperature with application to forehead
1 to 15 sec.
TempTrend™ II Temperature Indicators