Learn More
Biosynergy TempTrend™ Temperature Trend Indicators Available on GSA/VA Contract for Federal Government customers only.
Can be attached to all types of urine specimen containers
Manufacturer:  Biosynergy 1000B
 View more versions of this product
Catalog No. 22-246191
Add to cart
Description
Description
  • Disposable, liquid crystal temperature indicators
  • Single-spot readings in one to 15 seconds; temperature range from 87° to 103°F
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
87° to 103°F
87°F
Measuring temperature of urine specimens
Surface temperature indicator
103°F
Yes
1 to 15 sec.