Learn More
Biosynergy TempTrend™ Temperature Trend Indicators
Can be attached to all types of urine specimen containers
Manufacturer:  Biosynergy 1001B
 View more versions of this product
Catalog No. 22-267809
Add to cart
Description
Description
  • Disposable, liquid crystal temperature indicators
  • Single-spot readings in one to 15 seconds; temperature range from 87° to 103°F
Specifications
Specifications
87° to 103°F
87°F
Temperature and heat measurements
Temperature monitor
103°F
Yes
1 to 15 sec.