Learn More
Biosynergy TempTrend™ Temperature Trend Indicators Available on GSA/VA Contract for Federal Government customers only.
Can be attached to all types of urine specimen containers
$92.00 - $673.00
Specifications
Disposable Yes
Measurement Time 1 to 15 sec.
Temperature Range (English) 87°F to 103°F
View More Specs
Products 2
Catalog Number Mfr. No. Product Type For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type For Use With (Application) Price Quantity & Availability  
22-246191 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Biosynergy
1000B
Surface Temperature Indicator Measuring temperature of urine specimens Pack of 50 for $92.00
Case of 5 PK for $350.20
Add to cart
22-267809
View Documents
Biosynergy
1001B
Temperature Monitor Temperature and heat measurements Case of 1000 for $673.00
Add to cart
Description
Description
  • Disposable, liquid crystal temperature indicators
  • Single-spot readings in one to 15 seconds; temperature range from 87° to 103°F
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yes
1 to 15 sec.
87°F to 103°F