Learn More
Biotech Cheesemaking Products for Science Education
Manufacturer:  Neo/Sci Corporation 201453
Catalog No. S32630
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The ancient art of cheesemaking provides a perfect example of how biotechnology can be used to improve a process and its end-product.
  • Make cheese using enzymes produced through advances of biotechnology
  • Explore the science behind cheesemaking
  • Observe chemical and microbiological changes and the resulting physical changes
Additional investigations are included by changing such variables as temperature, pH, enzyme concentration, milk fat content and more. Class size: 40 students.
Specifications
Specifications
40 students
Explore the science behind cheesemaking and observe the accompanying chemical and microbiological changes and the resulting physical changes
Biotech Cheesemaking Lab Investigation Kit