Learn More
BioTek™ 405™ Touch Microplate Washer with Verify™ Technology Available on GSA/VA Contract for Federal Government customers only.
Features patented Verify™ technology which automatically checks for manifold tube blockage and visually reports any failed wells. Ultrasonic Advantage™ can be used to automatically and thoroughly clean the aspirate and dispense manifold.
$20960.00 - $32050.50
Specifications
Electrical Requirements 100 to 240VAC, 50/60Hz, ≤8.0 Ampere
Type Microplate Washer
View More Specs
Products 4
Catalog Number Mfr. No. Style Well Count Price Quantity & Availability  
Catalog Number Mfr. No. Style Well Count Price Quantity & Availability  
BT405TSRVSQ Available on GSA/VA Contract for Federal Government customers only.
View Documents
BioTek™
405TSRVSQ
Ultrasonic Advantage™, Verify™ technology, Biomagnetic separation, Vacuum filtration, 4-Buffer Switching 96 Each for $22,890.00
Add to cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
BT405TSUSQ Available on GSA/VA Contract for Federal Government customers only.
View Documents
BioTek™
405TSUSQ
Ultrasonic Advantage™, Verify™ technology, Biomagnetic separation, Vacuum filtration, Optimized Cell Washing 96 or 384 Each for $27,600.00
Add to cart
BT405TSRSQ Available on GSA/VA Contract for Federal Government customers only.
View Documents
BioTek™
405TSRSQ
Ultrasonic Advantage™, Verify™ technology, Biomagnetic separation, Vacuum filtration 96 Each for $20,960.00
Add to cart
BT405TSUVSQ Available on GSA/VA Contract for Federal Government customers only.
View Documents
BioTek™
405TSUVSQ
Ultrasonic Advantage™, Verify™ technology, Biomagnetic separation, Vacuum filtration, Optimized Cell Washing, 4-Buffer Switching 96 or 384 Each for $32,050.50
Add to cart
Description
Description
Hardware features and options:
  • Patented built-in Verify sensor automatically checks dispense / aspirate performance and reports clogged locations in the color graphical display.
  • Accommodates 96- and 384-well microplates
  • High resolution touch screen user interface
  • Onboard computer running Microsoft Windows CE
  • Patent pending built-in Ultrasonic Advantage automatically cleans clogged tubes
  • Two USB flash drive ports enable convenient file transfer, storage and operation
  • Automated internal 4-buffer switching
  • Internal positive displacement pump eliminates use of troublesome pressure delivery systems and allows use of any size dispense reservoir
  • Built-in priming trough allows automatic manifold priming and rinsing during both operation and maintenance modes and eliminates need for blank plates when used in automated robotic systems
  • All washers vacuum filtration ready (accessory filtration modules sold separately)
  • All washers biomagnetic separation ready (accessory magnets sold separately)
405 Select TS:
  • 96- or 384-well washing with patented 96-tube Dual-Action™ manifold
  • Cell washing is optimized with two low-flow rates and 20° angled dispense tubes ideal for washing loosely adherent cell monolayers
405 HT TS:
  • Fast 384-well washing with patented 192-tube Dual-Action™ manifold for independent dispense and aspiration control for precise overfill washing and overflow protection
  • Superior tube-in-tube aspiration manifold design for 384-well washing with buffers not containing surfactant
  • 96-well washing ready with optional 96-tube Dual-Action manifold
405 TS:
  • 405 TS models automate basic 96-well dispensing and aspiration
Software Features:
  • Context sensitive help system complete with recommendations, diagrams and images is only a key press away when questions arise
  • Pre-defined protocols and instructional videos walk users through instrument setup and basic use
  • Fully customizable protocol parameters for individualized wash requirements - gentle cell-washing to vigorous wash for high-binding assays
  • AutoPrime
  • Additional software flexibility and 21 CFR Part 11 compliance with Liquid Handling Control™ (LHC) Software for method development, downloading and washing plates.
  • Run protocols created onboard, downloaded from LHC™ Software or transferred on USB drive
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100 to 240VAC, 50/60Hz, ≤8.0 Ampere
Microplate Washer