BioTek™ MicroFlo™ Select Dispenser
Choose dynamic dispense volume, microplate types, tube sizes and dispense height
Manufacturer: BioTek™ 7170011
 View more versions of this product
Catalog No. 11-120-625
  Check Availability
Add to Cart
    Specifications
    Product Type Medium Volume Cassette
    Volume (Metric) Range 5 to 2500μL
    Diameter (English) 0.014 in.
    Diameter (Metric) 0.035cm
    For Use With (Application) For liquid handling
    For Use With (Equipment) For el406, multiflo™, multiflo™ fx
    Tip Material Molded Polypropylene
    Dispensing Increments 5μL
    • Purchase two BioTek™ MultiFlo™ FX/EL406 Dispenser Cassettes, and get one of equal or lesser value at no cost (up to $950 in savings). To take advantage of this offer, apply for the NLSU program.
      New Offer Buy Now
      Expires 2017-12-31
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback