BioTek™ MicroFlo™ Select Dispenser
Choose dynamic dispense volume, microplate types, tube sizes and dispense height
Manufacturer: BioTek™ 7170016
 View more versions of this product
Catalog No. 11-120-624
  Check Availability
Add to Cart
    Specifications
    Product Type Low Volume 1536 Well Cassette
    Diameter (English) 0.0075 in.
    Diameter (Metric) 0.019cm
    For Use With (Application) For liquid handling
    For Use With (Equipment) For el406, multiflo™, multiflo™ fx
    Tip Material Stainless Steel
    Dispensing Increments 1μL
    Volume (Metric) Range 1 to 50μL
    • Purchase two BioTek™ MultiFlo™ FX/EL406 Dispenser Cassettes, and get one of equal or lesser value at no cost (up to $950 in savings). To take advantage of this offer, apply for the NLSU program.
      New Offer Buy Now
      Expires 2017-12-31
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback