BioTek™ MicroFlo™ Select Dispenser
Choose dynamic dispense volume, microplate types, tube sizes and dispense height
Manufacturer: BioTek™ 7170014
 View more versions of this product
Catalog No. 11-120-626
  Check Availability
Add to Cart
    Specifications
    Product Type Medium Volume Cassette
    Diameter (English) 0.014 in.
    Diameter (Metric) 0.035cm
    For Use With (Application) For liquid handling
    For Use With (Equipment) For el406, multiflo™, multiflo™ fx
    Tip Material Stainless Steel
    Dispensing Increments 5μL
    Volume (Metric) Range 5 to 2500μL
  • Description & Specifications
Related Products
Product Content Feedback