Learn More
BioTek™ Synergy™ 2 Multi-Mode Microplate Readers Available on GSA/VA Contract for Federal Government customers only.
Designed for life science and drug discovery applications. BioTek Synergy 2 Multi-Mode Microplate Readers include dedicated optical elements which are optimized for each specific modality for low limits of detection in one compact instrument.
Manufacturer:  BioTek™ SLFPA
 View more versions of this product
Catalog No. 11-120-522
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Includes: Gen5™ software controlled through USB or serial port
Specifications
Description
  • Fluorescence intensity, time resolved fluorescence, fluorescence polarization, glow and flash luminescence, UV-Vis absorbance, FRET, TR-FRET, BRET, area scanning, adn spectral scanning
  • Optical system includes a narrow band monochromator, optimized filter/dichroic mirror pairing and three broad spectrum light sources — SQ Xenon Flash, Tungsten Halogen and High Energy DPR Xenon Flash
  • Modular design allows for easily upgrading elements
  • Optimized throughput for screening assays in 6- to 1536-well plates (luminescence 6-384) and PCR plates
  • Compatible with Take3 and Take3 Trio Micro-Volume plates — up to 16 or 48 2µL samples for low-volume DNA, RNA and protein quantification
  • Temperature control from 4° above ambient to 50°C
  • 11-120-515 includes two syringe pumps with dispensing volume of 5-1000µL in 1µL increments
Applications
  • For in vitro diagnostic use (except in the European Union)
  • Screening assays; Binding assays; Ion channel assays; Quantitative assays (DNA, protein); Kinetic assays; Gene expression assays (GFP, Luciferase); ELISA assays; Cell proliferation, Cytotoxicity
Compliance
CE and NRTL Safety Agency marked
Description & Specifications
Specifications
With fluorescence intensity and polarization, luminescence, UV-visible absorbance
0.001 O.D.
96 wells: 11 seconds; 384 wells: 22 seconds; 1536 wells: 42 seconds
17.5 in.
17 in.
14.5 in.
60 lb.
200 to 999nm; 1nm increments
0 to 4.0 O.D.
44.5cm
43.5cm
37.3cm
27kg