BioTek™ Synergy™ 2 Multi-Mode Microplate Readers Available on GSA/VA Contract for Federal Government customers only.

Designed for life science and drug discovery applications. BioTek Synergy 2 Multi-Mode Microplate Readers include dedicated optical elements which are optimized for each specific modality for low limits of detection in one compact instrument.

Manufacturer: BioTek™ BTSL

 View more versions of this product

Catalog No. 11-120-518

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

Includes: Gen5™ software controlled through USB or serial port

    Specifications

    Incubator Temperature 65°C
    Shaking Yes
    Packaging 1Each
    Software Type Gen5
    Model SLAD
    Product Type Multi Mode MicroPlate Reader
    Depth (English) 17.5 in.
    Width (Metric) 43.5cm
    Width (English) 17 in.
    Resolution 0.001 O.D.
    Dynamic Range 0 to 4.0 O.D.
    Measurement Speed 96 wells: 11 seconds; 384 wells: 22 seconds; 1536 wells: 42 seconds
    Depth (Metric) 44.5cm
    Height (Metric) 37.3cm
    Height (English) 14.5 in.
    Weight (Metric) 27kg
    Weight (English) 60 lb.
    Wavelength Range 200 to 999nm; 1nm increments

    • Fluorescence intensity, time resolved fluorescence, fluorescence polarization, glow and flash luminescence, UV-Vis absorbance, FRET, TR-FRET, BRET, area scanning, adn spectral scanning
    • Optical system includes a narrow band monochromator, optimized filter/dichroic mirror pairing and three broad spectrum light sources — SQ Xenon Flash, Tungsten Halogen and High Energy DPR Xenon Flash
    • Modular design allows for easily upgrading elements
    • Optimized throughput for screening assays in 6- to 1536-well plates (luminescence 6-384) and PCR plates
    • Compatible with Take3 and Take3 Trio Micro-Volume plates — up to 16 or 48 2µL samples for low-volume DNA, RNA and protein quantification
    • Temperature control from 4° above ambient to 50°C
    • 11-120-515 includes two syringe pumps with dispensing volume of 5-1000µL in 1µL increments

    Applications

    • For in vitro diagnostic use (except in the European Union)
    • Screening assays; Binding assays; Ion channel assays; Quantitative assays (DNA, protein); Kinetic assays; Gene expression assays (GFP, Luciferase); ELISA assays; Cell proliferation, Cytotoxicity

    Compliance

    CE and NRTL Safety Agency marked

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit