Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BioTek™ Synergy™ H1 Hybrid Multi-Mode All Detection Modes Microplate Readers Available on GSA/VA Contract for Federal Government customers only.

Flexible monochromator-based detection with high performance dichroic-based filter detection. BioTek Synergy H1 Hybrid Multi-Mode All Detection Modes Microplate Readers have monochromator optics which supports top and bottom fluorescence intensity, UV-visible absorbance and high-performance luminescence detection.

Manufacturer:  BioTek™ BTH1MFD

 View more versions of this product

Catalog No. 11-120-536


  Check Availability
Add to cart

Specifications

Description

  • Quadruple grating monochromator for maximum flexibility and ease of use
  • Compatible with Take3™ Multi-Volume Plate with 16 microspots of 2µL each for low-volume nucleic acid quantification
  • Dichroic-based filter optics for best performance and advanced detection technologies, such as fluorescence polarization and time-resolved fluorescence
  • Third generation quadruple grating design of the monochromator optics allows working at any excitation or emission wavelength with a 1nm step
  • Filter module includes its own light source and a high-performance dichroic-based wavelength selection system
  • Dual reagent injection system option automates ion channel assays and flash luminescent assays
  • Available with Gen5™ Data Analysis Software: reader control, advanced data analysis and flexible Excel export in one software package

Accessories

  • Take3 Multi-Volume Plate
  • BioStack™: 30 or 50 plate stacker
  • Gen5 Secure for 21CFR part 11 compliance
  • Fluorescence, Luminescence and Absorbance Test Plates

H1MFDG

  • Gas controller compatible
  • Gas module must be purchased separately (BT1210007 or BT12310008)

Note

  • Dual reagent dispenser option available with all models

Applications

  • Academic Life Science Research
  • Biotech industry—especially smaller companies and start-ups
  • Clinical—Filter module for emerging applications of TRF, FP and luminescence
Description & Specifications

Specifications

Microplate Reader; Dispenser
2.0
18.6 in.
15.4 in.
12.9 in.
50 lb.
Xenon flash
47.2cm
39.1cm
32.8cm
22.5kg
Monochromator fluorescence, full-light luminescence, filter/dichroic, time resolved fluorescence, polarization, filtered luminescence; Dual reagent dispenser
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit