BioTek™ Take3™ and Take3 Trio Micro-Volume Microplates Available on GSA/VA Contract for Federal Government customers only.
Take3 and Take3 Trio Micro-Volume plates expand functionality of the BioTek microplate spectrophotometers, multi-mode and Hybrid readers, while reducing sample volumes, costs and time required for routine quantification
Manufacturer: BioTek™ TAKE3TRIO
 View more versions of this product
Catalog No. 11-120-613
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Nucleic acid and protein quantification, in-situ fluorometric Pprotein quantification, 260/280 and 260/230 protein purity measurements, very low volume in-situ BCA assay, spectral scan of single or multiple low volume samples
    For Use With (Equipment) BioTek Eon™, Epoch™, Synergy™, PowerWave™ XS2 readers
    No. of Wells 48
    Volume (Metric) Well 2μL
    Applications Nucleic acid and protein quantification, in-situ fluorometric Pprotein quantification, 260/280 and 260/230 protein purity measurements, very low volume in-situ BCA assay, spectral scan of single or multiple low volume samples
    Wells 48
  • Description & Specifications
Related Products
Product Content Feedback