Learn More
Epredia™ Lab Vision™ Biotin Ab-2 Mouse Monoclonal Antibody, 200μg/mL with BSA and azide
Detect both the free and protein-conjugated forms of biotin with Epredia™ Biotin Ab-2, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS1048PABX
Description
Ab-1 has a very high affinity for biotin and is excellent for use in various amplification techniques. It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.Host Species: Mouse
Clone: BTN.4
Isotype: IgG1/κ
Species Reactivity: All species.
Immunogen: Biotin conjugated to BSA
Recommended for:
- Western Blotting
- Confocal Microscopy
- Flow Cytometry
- Image Analysis
- In situ Nucleic Acid Hybridization
Note:
USA: RUO; Int'l: RUO
Specifications
Biotin Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Epitope Tags | |
All species | |
IgG2a κ |
Western Blot | |
BTN.4 | |
Unconjugated | |
Biotin conjugated to BSA | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.