Learn More
Aat BioquestSupplier Diversity Partner Biotin Alkyne *CAS 1006592-45-5*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 3021
Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins. Biotin alkyne can be readily conjugated to many biomolecules through the well-known click chemistry.
Catalog No. 50-195-3209