Learn More
Biotin-Anti-His-Tag Antibody
Biotin-Anti-His-Tag Antibody
$1338.00 - $14290.00
Specifications
Concentration | 0.5 mg⁄ml |
---|---|
Molecular Weight of Antigen | 150kDa |
Packaging | 1 Each |
Primary or Secondary | Primary |
Content And Storage | Biotin-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C. |
Description
Biotin-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using biotin negatively impacts protein activity. These antibodies can be used with our Kinase Tracers and Eu-Streptavidin to conduct LanthaScreen™ Eu Kinase Binding Assays. Learn More and See Which Kinases Have Been Validated Using This AssaySpecifications
0.5 mg⁄ml | |
1 Each | |
Biotin-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C. |
150kDa | |
Primary | |
RUO |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.