Learn More
BiolegendSupplier Diversity Partner Biotin anti-human CD279 (PD-1)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 329934
Biotin anti-human CD279 (PD-1) [EH12.2H7]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Common Marmoset, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 ug
Catalog No. 50-712-053