Learn More
Pepro Tech Inc Biotinylated Anti-Human ApoA-I, 25ug DFS Item
Manufacturer:  Pepro Tech Inc 500P331BT25UG
Produced from sera of rabbits immunized with highly pure Recombinant Human ApoA-I. Anti-Human ApoA-I-specific antibody was purified by affinity chromatography and then biotinylated.
Catalog No. 50-195-2613