Learn More
Pepro Tech Inc Biotinylated Anti-Human sCD34, 1mg DFS Item
Manufacturer:  Pepro Tech Inc 500P327BT1MG
Produced from sera of rabbits immunized with highly pure Recombinant Human sCD34. Anti-Human sCD34-specific antibody was purified by affinity chromatography and then biotinylated.
Catalog No. 50-195-2594