Learn More
Pepro Tech Inc Biotinylated Anti-Human Uteroglobin, 25ug DFS Item
Manufacturer:  Pepro Tech Inc 500P330BT25UG
Produced from sera of rabbits immunized with highly pure Recombinant Human Uteroglobin. Anti-Human Uteroglobin-specific antibody was purified by affinity chromatography and then biotinylated.
Catalog No. 50-195-2607