Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin Antibody (BK-1/39), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250120AF700
Description
Biotin Monoclonal specifically detects Biotin in Human samples. It is validated for Immunohistochemistry.Specifications
Biotin | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
Cancer, Cellular Signaling, Epitope Tags, metabolism | |
Human | |
IgG1 |
Immunohistochemistry | |
BK-1/39 | |
Immunohistochemistry | |
Vitamin B4 | |
Biotin conjugated to keyhole limpet haemocyanin | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction