Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin Antibody (BK-1/39), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250120PCP
Description
Biotin Monoclonal antibody specifically detects Biotin in Human samples. It is validated for ImmunohistochemistrySpecifications
| Biotin | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Immunohistochemistry | |
| BK-1/39 | |
| Immunohistochemistry | |
| Vitamin B4 | |
| Biotin conjugated to keyhole limpet haemocyanin | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction