Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Biotin Deficient Diet, Powder

Diet deficient in biotin. Used in research.

$662.00 - $2530.00

Specifications

Type Animal Diet
Storage Requirements Room Temperature
Disposable Yes
Sterility Non-sterile
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP290103010
View Documents
MP Biomedicals
0290103010
10 kg
Each for $662.00
Add to cart
 
MP290103020
View Documents
MP Biomedicals
0290103020
20 kg
Each for $1,161.00
Add to cart
 
MP290103050
View Documents
MP Biomedicals
0290103050
50 kg
Each for $2,530.00
Add to cart
 
Description

Description

  • Diet deficient in biotin
  • Used in research
Specifications

Specifications

Animal Diet
Room Temperature
Yes
Non-sterile
SDS
Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit