Learn More
Aat BioquestSupplier Diversity Partner Biotin ethylenediamine *CAS 216299-38-6*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 3003
Biotin ethylenediamine is widely used to label biomolecules that contain either carboxy, phosphonyl or carbonyl group. For example, It is used for biotinylation of of nucleosides that have an aldehyde group.
Catalog No. 50-195-3196