Learn More
Vector LaboratoriesSupplier Diversity Partner Goat Anti-Rat IgG Antibody, Mouse Adsorbed Biotinylated 0.5 mg DFS Item
Manufacturer:  Vector LaboratoriesSupplier Diversity Partner BA9401
Vector Laboratories secondary antibodies are prepared using proprietary immunization schedules that produce high affinity antibodies. The antibodies are then purified by affinity chromatography, and cross-reactivities that are likely to interfere with specific labeling are removed by solid phase adsorption techniques. The biotinylated secondary antibodies are conjugated to ensure the maximum degree of labeling without compromising the specificity or affinity of the antibody. These antibodies are subjected to rigorous quality control assays and can be used for tissue and cell staining, ELISAs, and blots. Biotinylated Goat Anti-Rat IgG Antibody, Mouse Adsorbed, is supplied in solution and has been thoroughly adsorbed by a solid phase technique to remove antibodies that cross-react with mouse immunoglobulins. This adsorbed antibody allows primary antibodies made in goat against rat IgG to be used on mouse tissues. The recommended dilution for most applications is 1:100.
Catalog No. NC0113025