Learn More
Acro Biosystems BIOTIN HUM BCMA , FC TAG 25UG DFS Item
Manufacturer:  Acro Biosystems BC7H82F025UG
Biotinylated Human BCMA , Fc Tag (BC7-H82F0) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # NP_001183.2).
Catalog No. 50-113-8438