Learn More
Acro Biosystems Recombinant Protein;Biotinylated Human VEGF R2 / KDR Protein, Avi Tag (Avitag™);HEK293;25UG;KDR,KDR,CD309,FLK1,VEGFR,VEGFR2,VEGF R DFS Item
Manufacturer:  Acro Biosystems KDRH82E525UG
Recombinant Protein;Biotinylated Human VEGF R2 / KDR Protein, Avi Tag (Avitag™);25UG;Biotinylated Human VEGF R2, His Tag (KDR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 86 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R2, His Tag (Cat. No. KDR-H82E5) with a linear range of 0.15-2.5 μg/mL (QC tested).
Catalog No. 50-113-8416