Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin Hydrazide 98.0+%, TCI America™
SDP

Catalog No. p-7119671
Click to view available options
Quantity:
25 mg
100 mg

Chemical Identifiers

CAS 66640-86-6
Molecular Formula C10H18N4O2S
Molecular Weight (g/mol) 258.34
MDL Number MFCD00078532
InChI Key KOZWHQPRAOJMBN-ZKWXMUAHSA-N
Synonym biotin hydrazide, +-biotin hydrazide, 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanehydrazide, biotin-hydrazide, 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanehydrazi de, 5-3as,4s,6ar-2-oxo-1,3,3a,4,6,6a-hexahydrothieno 3,4-d imidazol-4-yl pentanehydrazide, biotin hydrazone, 5-3as,4s,6ar-2-oxo-hexahydrothieno 3,4-d imidazol-4-yl pentanehydrazide, biotin-nhnh2
PubChem CID 83872
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanehydrazide
SMILES C1C2C(C(S1)CCCCC(=O)NN)NC(=O)N2

Specifications

Melting Point 233°C
Color White
Quantity 25 mg
Formula Weight 258.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Biotin Hydrazide
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.