Learn More
Aat BioquestSupplier Diversity Partner Biotin-PEG3-azide *CAS 945633-30-7*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 3019
Biotin and biotin derivatives bind with high affinity to both avidins and streptavidins. This water-soluble biotin azide can be readily conjugated to many biomolecules through the well-known click chemistry.
Catalog No. 50-195-3207