Learn More
Aat BioquestSupplier Diversity Partner Biotin PEG4 succinimidyl ester DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 3022
Biotin PEG4 succinimidyl ester is an amine-reactive biotin derivative that contains a long arm to increase its avidin-binding affinity. This PEG biotin reacts specifically and efficiently with primary amines (e.g.
Catalog No. 50-195-3210