Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin, Powder, FCC, 97.5-100.5%, Spectrum™ Chemical
Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Biotin, Powder, FCC is often recommended as a dietary supplement for strenghening hair and nails. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Melting Range | 229-232°C |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Optical Rotation | +89° to +93° |
Percent Purity | 97.5 to 100.5% |
Grade | FCC |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction