Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin, Powder, FCC, 97.5-100.5%, Spectrum™

Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
58-85-5
C10H16N2O3S
244.31
YBJHBAHKTGYVGT-UHFFFAOYNA-N
5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
OC(=O)CCCCC1SCC2NC(=O)NC12
58-85-5
C10H16N2O3S
244.31
YBJHBAHKTGYVGT-UHFFFAOYNA-N
5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
OC(=O)CCCCC1SCC2NC(=O)NC12

BI115, 58-85-5, C10H16N2O3S

Spectrum™ Biotin, Powder, FCC is often recommended as a dietary supplement for strenghening hair and nails. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Optical Rotation +89° to +93°
Quantity 100 g
Melting Range 229-232°C
CAS Max % 1
Percent Purity 97.5 to 100.5%