Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin, Powder, USP, 97.5-102%, Spectrum™

Click to view available options
Quantity:
500 mg
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
58-85-5
C10H16N2O3S
244.31
YBJHBAHKTGYVGT-UHFFFAOYNA-N
5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
OC(=O)CCCCC1SCC2NC(=O)NC12
58-85-5
C10H16N2O3S
244.31
YBJHBAHKTGYVGT-UHFFFAOYNA-N
5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
OC(=O)CCCCC1SCC2NC(=O)NC12

B1103, 58-85-5, C10H16N2O3S

Spectrum™ Biotin, Powder, USP is used for preventing and treating biotin deficiency. Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Specific Rotation +89° to +93°
Quantity 1 g
CAS Max % 1
Percent Purity 97.5 to 102%