Learn More
Seracare Life Sciences IncSupplier Diversity Partner BIOTIN-RABBIT ANTI-LEGIONELLA  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 53600028
Protein G purified BacTrace antibody to Legionella species labeled with biotin. Isolated from a serum pool of rabbits immunized with various strains of heat-killed Legionella.
Catalog No. 50-672-91
Due to product restrictions, please to purchase or view availability for this product.