Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotin-Ubiquitin Non-distribution product as customer accommodation.

Biotin-Ubiquitin

$270.00

Specifications

Common Name Biotin-Ubiquitin
Concentration 1.19mg/mL
Formulation Frozen
Molecular Weight 9.2kDa
Name Biotin-Ubiquitin
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV4379 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV4379
10μg Each for $270.00
Add to cart
 
Description

Description

Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.

For use in combination with other LanthaScreen™ products including LanthaScreen™ Tb-Ubiquitin (PV4375), Fluorescein-Ubiquitin (PV4377), or a LanthaScreen™ antibody such as Tb-anti-biotin (PV4749).
Specifications

Specifications

Biotin-Ubiquitin
Frozen
Biotin-Ubiquitin
Biotin-Ubiquitin
–20°C
1.19mg/mL
9.2kDa
1 Each
For research use only. Not for use in diagnostic procedures
Frozen

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit