Learn More
Aat BioquestSupplier Diversity Partner Biotin-X NTA [Biotin-X nitrilotriacetic acid, potassium salt] *CAS 856661-92-2*  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 3009
Biotin-X nitrilotriacetic acid (biotin-X NTA) is widely used to detect histidine-tagged proteins immobilized on nitrocellulose membranes. The NTA moiety of biotin-X NTA chelates Ni ion that is also chelated with histidine tags.
Catalog No. 50-195-3201