Learn More
Biotinyl-[Glu1]-Gastrin I, Human, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66203MCR
 View more versions of this product
Catalog No. AAJ66203MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
2342.60
Human
Solid
RUO
Biotinyl-[Glu1]-Gastrin I, human
HPLC
1mg
Biotin-Glu-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2
-30° to -10°C