Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CD14 Protein, His,Avitag™
Supplier:  ACROBiosystems CD4H822825UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CD14, His,Avitag (CD4-H8228) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Met 344 (Accession # P08571-1 ).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 38.6 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6554