Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Latent TGF-beta 3 / Latent TGFB3 Protein, His,Avitag™
Supplier:  ACROBiosystems TG3H82E620UG
Encompass_Preferred
Recombinant Protein;20UG;Biotinylated Human Latent TGFB3, His,Avitag (TG3-H82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Ser 412 (Accession # P10600-1).;This protein carries a polyhistidine tag at the N-terminus and an Avi tag at the C-terminus. The protein has a calculated MW of 48.4 kDa. The protein migrates as 40-45 kDa and 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TGFBR2, Fc Tag (Cat. No. TG2-H5252) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGFB3, His,Avitag (Cat. No. TG3-H82E6) with a linear range of 0.3-20 ng/mL (QC tested).
Catalog No. 50-210-6929