Learn More
Acro Biosystems Recombinant Protein;Biotinylated Human LILRA2 / CD85h / ILT1 Protein, His Tag, Avi Tag (Avitag™);HEK293;LI2,ILT1,LIR7,CD85H,LIR-7,LILRA2,ILT-1,CD85h antigen,CD85 antigen-like family member H,leukocyte immunoglobulin-like receptor subfamily A member 2 DFS Item
Manufacturer:  Acro Biosystems LI2H82E925UG
Recombinant Protein;Biotinylated Human LILRA2 / CD85h / ILT1 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human LILRA2, His Tag, Avi Tag (LI2-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asn 449 (Accession # Q8N149-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.7 kDa. The protein migrates as 67-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRA2, His Tag, Avi Tag (Cat. No. LI2-H82E9) with a linear range of 0.625-5 μg/mL (QC tested).
Catalog No. NC1468905