Learn More
MBL International Corporation BIOTINYLATED MONOMER - HLA-A 0 DFS Item
Manufacturer:  MBL International Corporation TBM105M
Biotinylated Monomer - HLA-A*02:01 NY-ESO-1 C9V (SLLMWITQV) (formerly code #MR01076)
Catalog No. NC1464259