Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ Dulbecco's Modified Eagle's Medium with 4.5g L-Glucose per Liter, without L-Glutamine

Used in a wide range of mammalian cell culture applications.  BioWhittaker™ Dulbecco's Modified Eagle's Medium with 4.5g L-Glucose per Liter, without L-Glutamine is suitable for most types of cells, including human, monkey, hamster, rat, mouse, chicken and fish cellls and is manufactured in accordance with cGMP regulations.

Supplier:  Lonza™ 12614Q

 View more versions of this product

Catalog No. BW12-614Q

Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. View substitute products »

This item is not returnable. View return policy

Description

Description

  • DMEM is a modification of Basal Medium Eagle (BME)
  • Contains four-times the concentrations of the amino acids and vitamins
  • The high glucose version is well suited to high density suspension culture
  • Manufactured in accordance with cGMP regulations
  • The formulations used for standard liquid media products are those recommended by the Tissue Culture Association
Specifications

Specifications

2° to 8°C
DMEM Media
Sterile-filtered through two consecutive 0.2 μm filters
No Glutamine
10000 mL
Liquid
Sterile
Glucose
7 to 7.4
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit