Learn More
Lonza™ BioWhittaker™ RPMI 1640 with L-Glutamine
Developed for culturing normal human leukocytes. BioWhittaker RPMI 1640 with L-glutamine can be used for suspension and monolayer mammalian cultures.
Manufacturer:  Lonza™ 12115F/12
 View more versions of this product
Catalog No. BW12115F12
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Manufactured in accordance with cGMP regulations
  • Formulations used for standard liquid media products are those recommended by Tissue Culture Association
Specifications
Specifications
2° to 8°C
Sterile
Liquid
None
Sterile-filtered through two consecutive 0.2μm pharmaceutical-grade filters
2° to 8°C
12 × 500mL
w/ L-Glutamine and 25mM HEPES Buffer
7.12 to 7.48
RPMI 1640