Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biperiden hydrochloride, Tocris Bioscience™

Catalog Number p-4783798
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
1235-82-1
C21H30ClNO
347.927
RDNLAULGBSQZMP-LIJKWCKLSA-N
1r-1-1r,4r,5s-5-bicyclo 2.2.1 hept-2-enyl-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
129318274
(1R)-1-[(1R,4R,5S)-5-bicyclo[2.2.1]hept-2-enyl]-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
C1CCN(CC1)CCC(C2CC3CC2C=C3)(C4=CC=CC=C4)O.Cl
This item is not returnable. View return policy
1235-82-1
C21H30ClNO
347.927
RDNLAULGBSQZMP-LIJKWCKLSA-N
1r-1-1r,4r,5s-5-bicyclo 2.2.1 hept-2-enyl-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
129318274
(1R)-1-[(1R,4R,5S)-5-bicyclo[2.2.1]hept-2-enyl]-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
C1CCN(CC1)CCC(C2CC3CC2C=C3)(C4=CC=CC=C4)O.Cl

Muscarinic receptor antagonist; some selectivity for M1

Muscarinic receptor antagonist that displays some selectivity for the M1 subtype (Ki values are 0.48, 2.4, 3.9, 6.3 and 6.3 nM for M1, M4, M3, M2 and M5 receptors respectively). Alleviates extrapyramidal symptoms associated with antipsychotic drugs. Displays antiparkinsonian activity.
Quantity 10 mg
Formula Weight 347.92
Percent Purity >99%
Chemical Name or Material Biperiden hydrochloride
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit