Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Biphenyl-2-boronic acid pinacol ester, 97%

Manufacturer:  Alfa Aesar™ H3232014

Quantity 25g

 View more versions of this product

Catalog No. AAH3232014


  Check Availability
Add to cart

Specifications

Description

Applications
Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Description & Specifications

Chemical Identifiers

914675-52-8
280.17
Biphenyl-2-boronic acid pinacol ester
C18H21BO2
MFCD14636459

Specifications

914675-52-8
280.17
76° to 79°C
Biphenyl-2-boronic acid pinacol ester
C18H21BO2
MFCD14636459
25g
97%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit