Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Biphenyl-4-boronic acid, 98+%

Click to view available options
Quantity:
1g
5g
25g
5122-94-1
C12H11BO2
198.028
MFCD00093311
XPEIJWZLPWNNOK-UHFFFAOYSA-N
4-biphenylboronic acid,1,1'-biphenyl-4-ylboronic acid,biphenyl-4-boronic acid,4-biphenyl boronic acid,biphenylboronic acid,4-phenylphenyl boranediol,4-phenylphenyl boronic acid,biphenyl-4-ylboronic acid,1,1'-biphenyl-4-yl-boronic acid,4-phenylbenzeneboronic acid
151253
(4-phenylphenyl)boronic acid
B(C1=CC=C(C=C1)C2=CC=CC=C2)(O)O
5122-94-1
C12H11BO2
198.028
MFCD00093311
XPEIJWZLPWNNOK-UHFFFAOYSA-N
4-biphenylboronic acid,1,1'-biphenyl-4-ylboronic acid,biphenyl-4-boronic acid,4-biphenyl boronic acid,biphenylboronic acid,4-phenylphenyl boranediol,4-phenylphenyl boronic acid,biphenyl-4-ylboronic acid,1,1'-biphenyl-4-yl-boronic acid,4-phenylbenzeneboronic acid
151253
(4-phenylphenyl)boronic acid
B(C1=CC=C(C=C1)C2=CC=CC=C2)(O)O
Applications
Used in Suzuki reactions and as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Odor Odorless
Quantity 25g
Melting Point 238°C to 247°C
Beilstein 2937410
Formula Weight 198.03
Percent Purity ≥98%
Chemical Name or Material Biphenyl-4-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning