Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biphenyl, 99%, Spectrum™

Click to view available options
Quantity:
25 g
250 g
1 kg
20 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
92-52-4
92-52-4
Packaging Poly Bottle
Quantity 250 g
CAS Max % 1
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.