Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BirA Mouse anti-E. coli, Janelia Fluor 549, Clone: 5B11c3-3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259939JF549

Catalog No. NBP59939549


Add to cart

Description

Description

BirA Monoclonal antibody specifically detects BirA in E. coli samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

BirA
5B11c3-3
Immunocytochemistry/Immunofluorescence
Purified
birA
GST fused to E.coli BirA R118G
Protein A purified
Store at 4°C in the dark.
Monoclonal
The antibody recognises the C-terminal half of E.coli BirA; the antibody recognised the BirA variant miniTurbo; not tested for TurboID or other BirA derivate.
Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Bifunctional protein birA, Bio R, BioR, Biotin [acetyl CoA carboxylase] synthetase
Mouse
IgG1 Kappa
0.1mL
Primary
948469
E. coli
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit