Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIRC7 Mouse anti-Human, Clone: 88C570, Abnova™

Mouse monoclonal antibody raised against recombinant human BIRC7.

Manufacturer:  Abnova Corporation MAB12961

Catalog No. 89-933-643


Add to cart

Description

Description

The protein encoded by this gene is a member of the family of inhibitor of apoptosis proteins (IAP) and contains a single copy of a baculovirus IAP repeat (BIR) as well as a RING-type zinc finger domain. The BIR domain is essential for inhibitory activity and interacts with caspases, while the RING finger domain sometimes enhances antiapoptotic activity but does not inhibit apoptosis alone. Two transcript variants encoding different isoforms have been found for this gene. The two isoforms have different antiapoptotic properties, with isoform alpha protecting cells from apoptosis induced by staurosporine and isoform b protecting cells from apoptosis induced by etoposide. [provided by RefSeq]

Specifications

Specifications

BIRC7
88C570
Mouse monoclonal antibody raised against recombinant human BIRC7.
In PBS (0.05% BSA, 0.05% sodium azide).
KIAP/LIVIN/ML-IAP/MLIAP/RNF50
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
BIRC7
Recombinant protein corresponding to human BIRC7.
Protein G purification
RUO
Primary
79444
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit