Learn More
BIRC8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 201441AP
Catalog No. 50-173-2730
Add to cart
Description
Description
Apoptosis, or programmed cell death, is related to many diseases, such as cancer. Apoptosis is triggered by a variety of stimuli including members in the TNF family and prevented by the inhibitor of apoptosis (IAP) proteins. IAP proteins form a conserved gene family including IAP, XIAP/ILP-1/MIHA, and Livin/KIAP that bind to and inhibits specific proteases. A novel member in the IAP protein family was recently identified and designated ILP-2 for IAP-like protein-2. ILP-2 has high homology to ILP-1, but is encoded by a distinct gene that is solely expressed in testis of tested normal human tissues. ILP-2, unlike ILP-1, has no inhibitory effect on Fas and TNF induced apoptosis, but potently inhibits apoptosis induced by overexpression of Bax or by coexpression of caspase-9 with Apaf-1. ILP-2 interacts with the processed caspase-9. These results suggest that ILP-2 is a novel IAP family member with restricted specificity for caspase-9.
Specifications
Specifications
BIRC8
0.53 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q60989, Q96P09
BIRC8, hILP2, IAP like protein 2, ILP 2, ILP2
Rabbit
IgG
150 μL
-20°C
Primary
112401, 11798
Western Blot
Unconjugated
BIRC8
Liquid
BIRC8, Xiap
BIRC8 Fusion Protein Ag13658
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse